Order Entry
Austria
ContactUsLinkComponent
 
Anti-FBXO7 Rabbit Polyclonal Antibody
  AVIVARP43128_P050
Anti-FBXO7 Rabbit Polyclonal Antibody
  AVIVARP43128_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    F-box protein 7
  • Antigen symbol:
    FBXO7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    25793
  • Antigen synonyms:
    PKPS|FBX07|FBX7|PARK15|FBX
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: FBXO7 (F-box protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: