Anti-PPP2R3B Rabbit Polyclonal Antibody
Catalog # AVIVARP42371_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:protein phosphatase 2, regulatory subunit B'', beta
- Antigen symbol:PPP2R3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:28227
- Antigen synonyms:PR48|NYREN8|PPP2R3LY|PPP2R3L
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PPP2R3B (protein phosphatase 2, regulatory subunit B'', beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: