Anti-SIGLEC6 Rabbit Polyclonal Antibody
Catalog # AVIVARP41925_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 6
- Antigen symbol:SIGLEC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:946
- Antigen synonyms:CD33L|CD327|OBBP1|CD33L2|CDW327|CD33L1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SIGLEC6 (sialic acid binding Ig-like lectin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human