Anti-UGT2B15 Rabbit Polyclonal Antibody
Catalog # AVIVARP41906_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:UDP glucuronosyltransferase 2 family, polypeptide B15
- Antigen symbol:UGT2B15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7366
- Antigen synonyms:UDPGT2B15|HLUG4|UGT2B8|UDPGTH3|UDPGT 2B8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: UGT2B15 (UDP glucuronosyltransferase 2 family, polypeptide B15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human