Anti-HNRNPC Rabbit Polyclonal Antibody
Artikelnr. AVIVARP40517_P050
Lieferant: Aviva Systems Biology
Spezifikationen
- Antibody type:Primary
- Antigen name:Heterogeneous nuclear ribonucleoprotein C
- Antigen symbol:HNRNPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:3183
- Antigen synonyms:C1|HNRNP|SNRPC|HNRPC|C2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: HNRNPC (heterogeneous nuclear ribonucleoprotein C (C1/C2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: