Order Entry
Austria
ContactUsLinkComponent
Anti-TUT1 Rabbit Polyclonal Antibody
Anti-TUT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP39566_P050
Anti-TUT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP39566_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Terminal Uridylyl Transferase 1 U6 SnRNA-specific
  • Antigen symbol:
    TUT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    499314
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: TUT1 (terminal uridylyl transferase 1, U6 snRNA-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat