Anti-HSFX1 Rabbit Polyclonal Antibody
Catalog # AVIVARP35906_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Heat shock transcription factor family, X linked 1
- Antigen symbol:HSFX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:51402
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HSFX1 (heat shock transcription factor family, X linked 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human