Order Entry
Austria
ContactUsLinkComponent
Anti-CCNDBP1 Rabbit Polyclonal Antibody
Anti-CCNDBP1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7885R
Supplier:  Bioss
Anti-CCNDBP1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7885R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin D-type Binding-protein 1
  • Antigen symbol:
    CCNDBP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    GCIP|grap2 and cyclin-D-interacting protein|DIP1|human homolog of Maid|cyclin-D1-binding protein 1|cyclin D-type binding-protein 1|HHM
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CCNDBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat