Anti-PGRPS Rabbit Polyclonal Antibody
Catalog # BOSSBS-7673R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peptidoglycan Recognition Protein 1
- Antigen symbol:PGRPS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:PGLYRP|TAG7|member 3 (LTB)-like (peptidoglycan recognition protein)|PGLYRP1|PGRP|MGC126896|TNFSF3L|peptidoglycan recognition protein 1|TNF superfamily|PGRP-S|PGRPS|MGC126894
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PGRPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat