Anti-AKT3 Rabbit Polyclonal Antibody
BOSSBS-5209R
: Bioss
- Antibody type:Primary
- Antigen name:V-Akt Murine Thymoma Viral Oncogene homolog 3
- Antigen symbol:AKT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:PKB-GAMMA|PKBG|RAC-PK-gamma|STK-2|RAC-gamma|PRKBG|MPPH2|MPPH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat