Anti-TXNIP Rabbit Polyclonal Antibody
Catalog # BOSSBS-3897R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Thioredoxin-interacting protein
- Antigen symbol:TXNIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:EST01027|BMP and activin membrane-bound inhibitor homolog|HHCPA78|THIF|VDUP1|NMA|non-metastatic gene A protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TXNIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat