Anti-ID1 Rabbit Polyclonal Antibody
BOSSBS-3529R
: Bioss
- Antibody type:Primary
- Antigen name:Inhibitor Of Differentiation 1
- Antigen symbol:ID1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:BHLHb24|Dominant Negative Helix-Loop-Helix Protein|Class B Basic Helix-Loop-Helix Protein 24|DNA-Binding Protein Inhibitor ID-1|Inhibitor Of DNA Binding 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat