Anti-RELA Rabbit Polyclonal Antibody
BOSSBS-3485R
: Bioss
- Antibody type:Primary
- Antigen name:Transcription factor p65
- Antigen symbol:RELA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:nuclear factor NF-kappa-B p65 subunit|v-rel reticuloendotheliosis viral oncogene homolog A|p65|nuclear factor of kappa light polypeptide gene enhancer in B-cells 3|transcription factor p65|v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 (p65))
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat