Order Entry
Austria
ContactUsLinkComponent
 
Anti-YB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3477R
Supplier:  Bioss
undefined
Anti-YB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3477R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Y box binding Protein 1
  • Antigen symbol:
    YB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    CCAAT-binding transcription factor I subunit A|EFI-A|YB1|DBPB|CBF-A|Y-box-binding protein 1|NSEP-1|nuclease-sensitive element-binding protein 1|CSDA2|Y box-binding protein I|DNA-binding protein B|major histocompatibility complex|NSEP1|MDR-NF1|class II|BP-8|MGC110976|enhancer factor I subunit A|MGC117250|CSDB|nuclease sensitive element binding protein 1|YB-1|MGC104858|Y-box transcription factor
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: YB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat