Anti-COL11A2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2924R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Collagen, type XI, alpha 2
- Antigen symbol:COL11A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:DFNA13|FBCG2|DFNB53|STL3|HKE5|PARP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: COL11A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: