Anti-CD14 Rabbit Polyclonal Antibody
Artikelnr. BOSSBS-1192R
Lieferant: Bioss
Spezifikationen
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Antigen symbol:CD14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat