Order Entry
Austria
ContactUsLinkComponent
Anti-CTNNB1 Rabbit Polyclonal Antibody
Anti-CTNNB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1165R
Supplier:  Bioss
undefined
Anti-CTNNB1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1165R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Catenin beta
  • Antigen symbol:
    CTNNB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Antigen synonyms:
    Beta-catenin|ORF Names: OK/SW-cl.35|Catenin beta-1|PRO2286|Catenin beta|CTNNB|CTNNB1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: