Anti-CDH1 Rabbit Polyclonal Antibody
BOSSBS-10009R
: Bioss
- Antibody type:Primary
- Antigen name:Cadherin 1
- Antigen symbol:CDH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: