Order Entry
Austria
ContactUsLinkComponent
Anti-HPV16 E6 protein Rabbit Polyclonal Antibody
Anti-HPV16 E6 protein Rabbit Polyclonal Antibody
Catalog # BOSSBS-0990R
Supplier:  Bioss
undefined
Anti-HPV16 E6 protein Rabbit Polyclonal Antibody
Catalog # BOSSBS-0990R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HPV16 E6 Protein
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: HPV16 E6 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus