Order Entry
Austria
ContactUsLinkComponent
Combined high speed plate/microtubeshaker/vortex mixer, MPS-1
Combined high speed plate/microtubeshaker/vortex mixer, MPS-1
Supplier:  GRANT INSTRUMENTS
Certificates
undefined
Combined high speed plate/microtubeshaker/vortex mixer, MPS-1
Supplier:  GRANT INSTRUMENTS

Specifications

  • Catalog No:
  • 444-0961
  • 444-0962
  • Model:
  • MPS-1
  • MPS-1
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbit:
  • 3 mm
  • 3 mm
  • Speed range:
  • 300 - 3200 min⁻¹
  • 300 - 3200 min⁻¹
  • Max. capacity:
  • 50 ml
  • 50 ml
  • W×D×H:
  • 234×220×150 mm
  • 234×220×150 mm
  • Weight:
  • 4,7 kg
  • 4,7 kg

Specifications

Frequently Bought Together