Sodium hydroxide 0.1 mol/L in aqueous solution, EMPROVE® EXPERT for biopharmaceutical production
Catalog # 1.37058.9950
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pk:950 L
- Seq:5783
Specifications
Specification Test Results
- Assay (acidimetric,NaOH):0.05- 0.15 mol/l
- Carbonate (as Na2CO3):≤ 0.2%
- Chloride (Cl):≤ 5%
- Sulfate (SO.):≤ 30 ppm
- Heavy metals (as Pb):≤ 5.0 ppm
- Al (Aluminium):≤ 10 ppm
- Fe (Iron):≤ 3 ppm
- Residual solvents (ICH Q3C):excluded by production process
- Endotoxins:≤ 2.0 I.U./ml
Specification Test Results
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,
:Filtered through a 0.2 µm filter.
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548