Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Supplier:   SciLabware
Multiple adapters, Quickfit®
Supplier:  SciLabware
Certificates
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two necks. One vertical and one angled at 45°.
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,20 €
    60,20 €
    1 * 1 ST
       60,20 €
  • 24/29 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,30 €
    58,30 €
    1 * 1 ST
       58,30 €