ContactUsLinkComponent
Wafer shipping system, horizontal, SmartStack®
Catalog # ENTEHWS-KIT-CUSHE
Supplier:  ENTEGRIS
Wafer shipping system, horizontal, SmartStack®
Catalog # ENTEHWS-KIT-CUSHE
Supplier:  ENTEGRIS
Specifications
  • Description:
    Outer cushion for SmartStack® 1-pack
  • Seq:
    02
  • Pk:
    100
Specifications
About this item
SmartStack® horizontal wafer shipping system is designed to reduce wafer breakage, die bond corrosion and particle contamination. These shipping system is designed to utilize common secondary packaging across wafer sizes, and the result is reduction in the number of pacvkaging configurations and wafer packing process. With additional layer of protection it is easy to maintain wafer integrity during transport and shipping.
  • Multiple configurations are common across wafer sizes as 1-pack system holds one canister and 2-pack hold two canisters
  • Passes ISTA-2A drop test
  • Accommodates 150 and 200 mm SmartStack® canisters
Ordering information: The system includes corrugated ship box, outer packaging cushion and ESD bags but should be ordered separately.