Order Entry
Austria
ContactUsLinkComponent
VWR® Q Path® Labofix Cytology Fixative
VWR® Q Path® Labofix Cytology Fixative
Supplier:  VWR Chemicals
Certificates
undefined
VWR® Q Path® Labofix Cytology Fixative
Supplier:  VWR Chemicals
Certificates

Specifications

  • Form:
    Colourless liquid
  • Density:
    0,820 – 0,860

Specifications

Frequently Bought Together