Inline equilibrium cell, SP Bel-Art
Catalog #
Product Details & Documents
Clear acrylic construction with seven stainless steel screws and wing nuts for clamping cells and dialysis membrane. The cavity plug screws are Teflon® TFE coated stainless steel.
- Each half cavity has a top opening for introduction or withdrawal of solutions and holds 1 ml in each half cell