Orders LinkContactUsLinkComponent
Container, Securitainer®
Container, Securitainer®
Supplier:   COVERA
Container, Securitainer®
Supplier:  COVERA
Certificates
Container, Securitainer®
Container, Securitainer®
14 Options Available Below
Product Details & Documents
SECURITAINER®, made of white PP is an effective security container. It has two main uses: as an individual transport container for glass vials and as packaging for materials and substances sensitive to dampness.
  • The tamper-proof seal guarantees that the packaging has not been opened-providing increased product security
  • The seal prevents manipulation of vials, labels and their contents
  • The strong, light construction protects the contents effectively
  • Easy to handle
  • Lids to be ordered seperately
Product Family Options
Product Information
  • Capacity
    Height
    Cap Size
    Neck-Ø
    Availability
    Price
  • 27 ml
    51 mm
    26 mm
    26 mm
    Log in for availability
    Log in for availability
    1 * 1.250 ST
    1 * 1.250 ST - 510,00 €
    510,00 €
    1 * 1.250 ST
       510,00 €
  • 49 ml
    52 mm
    35 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 700 ST
    1 * 700 ST - 334,00 €
    334,00 €
    1 * 700 ST
       334,00 €
  • 77 ml
    82 mm
    35 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 200,00 €
    200,00 €
    1 * 400 ST
       200,00 €
  • 105 ml
    58 mm
    49 mm
    49 mm
    Log in for availability
    Log in for availability
    1 * 300 ST
    1 * 300 ST - 252,00 €
    252,00 €
    1 * 300 ST
       252,00 €
  • 135 ml
    75 mm
    49 mm
    49 mm
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 205,00 €
    205,00 €
    1 * 240 ST
       205,00 €
  • 153 ml
    85 mm
    49 mm
    49 mm
    Log in for availability
    Log in for availability
    1 * 220 ST
    1 * 220 ST - 260,00 €
    260,00 €
    1 * 220 ST
       260,00 €
  • 260 ml
    90 mm
    62 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 135 ST
    1 * 135 ST - 219,00 €
    219,00 €
    1 * 135 ST
       219,00 €
  • 327 ml
    113 mm
    62 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 320,00 €
    320,00 €
    1 * 180 ST
       320,00 €
  • 880 ml
    100 mm
    109 mm
    109 mm
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 240,00 €
    240,00 €
    1 * 48 ST
       240,00 €
Securitainer®-Caps
Product Information
  • Capacity
    Height
    Cap Size
    Neck-Ø
    Availability
    Price
  • COVEC26/01
    Securitainer®-cap
    Securitainer®-cap
    26 mm
    Log in for availability
    Log in for availability
    1 * 3.900 ST
    1 * 3.900 ST - 631,00 €
    631,00 €
    1 * 3.900 ST
       631,00 €
  • COVEC35/01
    Securitainer®-cap
    Securitainer®-cap
    35 mm
    Log in for availability
    Log in for availability
    1 * 1.675 ST
    1 * 1.675 ST - 665,00 €
    665,00 €
    1 * 1.675 ST
       665,00 €
  • COVEC49/01
    Securitainer®-cap
    Securitainer®-cap
    49 mm
    Log in for availability
    Log in for availability
    1 * 1.200 ST
    1 * 1.200 ST - 554,00 €
    554,00 €
    1 * 1.200 ST
       554,00 €
  • COVEC62/01
    Securitainer®-cap
    Securitainer®-cap
    62 mm
    Log in for availability
    Log in for availability
    1 * 900 ST
    1 * 900 ST - 626,00 €
    626,00 €
    1 * 900 ST
       626,00 €
  • COVEC109/01
    Securitainer®-cap
    Securitainer®-cap
    109 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 388,00 €
    388,00 €
    1 * 180 ST
       388,00 €