ContactUsLinkComponent
Handheld digital microscope, Dino-Lite Medical, Podoscope
Handheld digital microscope, Dino-Lite Medical, Podoscope
Catalog # 630-2611
undefined
Handheld digital microscope, Dino-Lite Medical, Podoscope
Catalog # 630-2611
Specifications
  • Magnification:
    10 - 50×
  • Model:
    PodoScope (MEDL4FW)
  • Resolution:
    1,3 MP
  • Description:
    Handheld microscope with CMOS sensor
  • Additional features:
    Anti-reflection polariser
  • Illumination:
    8 LEDs
  • Pk:
    1
Specifications
About this item
With this handheld microscope changes in the skin of the foot and the foot itself can be documented by taking photographs and making movies as well. The podiatrist and his patient can view the images in real time on a laptop, Windows tablet or computer.
  • Versatility due to exchangeable caps
  • Built-in lighting (eight bright white LEDs)
  • High speed USB 2.0 connection
  • Powered via USB port: No batteries needed
  • Built-in anti-reflection polariser will help to reduce any bothersome shine or glow of the skin
Designed for use by podiatrists.
Certifications: Certified according to the Medical Device Directive 93/42/EEC, modified 2007/47/EC.
Delivery: Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), calibration target and medical user manual.