Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps for laboratory bottles
Screw caps for laboratory bottles
MARI5444722
:
MARIENFELD
Screw caps for laboratory bottles
MARI5444722
:
MARIENFELD
Thread:
GL 45
Colour:
Blue
Description:
Screw cap, PP
Pk:
10