Accessories for multi-parameter photometers
Supplier: Palintest
Certificates
About this item
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Orthophosphoric acid 85% 85% ≥85%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
Hydrometers for low and high surface tension
Supplier: Alla France
Guanidinium thiocyanate ≥99.0% for biotechnology
Supplier: VWR Chemicals