ContactUsLinkComponent
Cold traps, type KF NW
Cold traps, type KF NW
Catalog # KGWA17380-NW16
Supplier:  KGW
undefined
Cold traps, type KF NW
Catalog # KGWA17380-NW16
Supplier:  KGW
Specifications
  • Description:
    Cold trap with NW flange, long version, KFL 29-NW 16-A, with spout, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 ml
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Simple cold traps, which are also known as cold finger condensers, are used in conjunction with vacuum pumps and the condensation Produced from humidity or solvents these cold traps can be used for many different tasks. With glass vacuum flanges you are able to connect this cold trap directly to the metal flanges of a high vacuum pump.
Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Reliable, perfect version, easy to clean and handle
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminum
  • Pressure-free cooling sphere inside the Dewar flask
These simple cold finger condensers are used in standard applications in conjunction with vacuum pumps and the condensation is forced out of the humidity or solvents used in the chemical applications. Cold traps can be changed very quickly without any problems and it can be connected directly to the metal flanges of a high vacuum pump with its glass vacuum flanges.