ContactUsLinkComponent
Peqlab, PCR Plates, 384-Well, Skirted
Catalog # 732-3237
Supplier:  VWR Peqlab
   Low Manufacturing Impact
undefined
Peqlab, PCR Plates, 384-Well, Skirted
Catalog # 732-3237
Supplier:  VWR Peqlab
   Low Manufacturing Impact
Specifications
  • No. of wells:
    384
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Seq:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    50
  • Well volume:
    0,04 ml
Specifications
About this item
PP, autoclavable. These high quality, 384-well PCR plates are suitable for high throughput applications, and are designed to be compatible with robotic platforms and automated liquid handling systems.
  • Compatible with most 384-well thermal cycler systems and robots (colour-free plates)
  • 40 µl maximum well capacity and 25 µl operating volume
  • Cut away corner top right and bottom right for easy orientation
  • Manufactured from virgin polypropylene under strict quality control guidelines
  • Certified free from RNases, DNases and human DNA