ContactUsLinkComponent
Peqlab, PCR Plates, 96-Well, Skirted, Low Profile
Peqlab, PCR Plates, 96-Well, Skirted, Low Profile
  732-3225
   Low Manufacturing Impact
undefined
Peqlab, PCR Plates, 96-Well, Skirted, Low Profile
  732-3225
   Low Manufacturing Impact
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    25
PP. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation for enhanced reaction reproducibility. Full skirt gives greater stability.
  • Ultra-thin, low profile design for effective heat transfer and minimum condensation
  • Suitable for use with automated systems
  • Eight holes in skirt aid plate positioning and removal from thermal cycler block
  • Cut-away corner (H1)
  • Certified free from DNase, RNase and human genomic DNA