Peqlab, PCR Plates, 96-Well, Low Profile
Supplier: VWR Peqlab
Certificates
About this item
PP, non-skirted. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation for enhanced reaction reproducibility.
- Effective design with four overhanging edges for easy handling
- Alphanumeric coding for quick sample identification
- Certified free from RNase, DNase and human DNA
- White plates are ideal for use in Real-Time PCR
- Compatible with a wide range of thermal cyclers
- Cut away corner (H12)
Specifications
- Catalog No:
- 732-3220
- 732-3221
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)