Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
EDTA disodium salt concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84864.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA disodium salt concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84864.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    170 mL
  • Seq:
    2515

Specifications

Specification Test Results

  • Titer (20°C):
    0.0995 - 0.1005 mol/l

Specification Test Results

About this item

 :
 :336.21 g/mol
 :Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672