ContactUsLinkComponent
PIC (paired ion chromatography) reagents for separation by reverse phase chromatography
PIC (paired ion chromatography) reagents for separation by reverse phase chromatography
Catalog # WATEWAT085103
Supplier:  WATERS
CAS Number:  
PIC (paired ion chromatography) reagents for separation by reverse phase chromatography
Catalog # WATEWAT085103
Supplier:  WATERS
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Paired-ionic chromatography reagent B7, heptane sulfonic acid