Order Entry
Austria
ContactUsLinkComponent
Peqlab PerfectBlue Maxi M 'Revolution', Gel system
Peqlab PerfectBlue Maxi M 'Revolution', Gel system
Catalog # 700-0435
Supplier:  VWR Peqlab
undefined
Peqlab PerfectBlue Maxi M 'Revolution', Gel system
Catalog # 700-0435
Supplier:  VWR Peqlab

Specifications

  • Description:
    PerfectBlue™ Gel System Maxi M 'Revolution'
  • Seq:
    01
  • Model:
    Maxi M 'Revolution'
  • Gel volume:
    250 ml
  • Buffer volume:
    ~2000 ml
  • Run time:
    ≥60 h
  • Gel size W×L:
    200×250 mm
  • Sample capacity:
    8 - 168
  • Unit W×L×H:
    270×380×155 mm
  • Pk:
    1

Specifications

About this item

Based on the Maxi M model, the Maxi M 'Revolution' offers an in-built buffer circulation system, powered by the system's electrodes. At the cathode, hydrogen bubbles are produced and pass up a diagonally positioned pipe connected to the two sides of the tank to produce a gentle and continual circulation of the buffer. The system nulls any pH and ionic gradient – without the fuss of mechanical pumps. The system offers 6 comb positions enabling the gel to be partitioned into 2, 3 or 4 areas. Additionally, the system can accommodate any of the 23 standard and microtitre comb designs described for the Maxi M model.

*Based on an agarose gel of approximately 5 mm thickness

Ordering information: Supplied with 3 combs (1,5 mm thickness, 16, 24, 36 teeth), lid, gel tray, 3-point levelling platform, 'end gates', tank and cables.