ContactUsLinkComponent
Racks for centrifuge tubes, SP Bel-Art
Racks for centrifuge tubes, SP Bel-Art
Catalog # BELAF187960000
undefined
Racks for centrifuge tubes, SP Bel-Art
Catalog # BELAF187960000
Specifications
  • For tubes:
    15/50 ml
  • No. of Holes:
    1
  • Pk:
    4
Specifications
About this item
PP, autoclavable
  • Supports conical bottom tubes
  • Open uprights for clear view of entire contents of the tube