ContactUsLinkComponent
Spectrophotometric colorimeters, model AF, Lovibond®
Spectrophotometric colorimeters, model AF, Lovibond®
Catalog # TINT107103
undefined
Spectrophotometric colorimeters, model AF, Lovibond®
Catalog # TINT107103
Specifications
  • Light source:
    2×12 Volt, 20 W tungsten halogen lamp
  • Description:
    AF710-3
  • Model:
    Model AF
  • Illuminants:
    Approximates to daylight
  • Optical design:
    Visual colour comparison against colour glasses
  • Weight:
    8,3 kg
  • W×D×H:
    330×100×480 mm
  • Pk:
    1
  • For:
    Oils and fats
Specifications
About this item
The AF710-3 is a visual instrument used for colour grading of oils and fats.
Colour is determined by comparing light transmitted through a column of oil with that transmitted through a series of Red and Yellow glasses calibrated in accordance with the AOCS-Tintometer® Colour Scale.
The glasses enable a colour match to be found from the light passing through the oil. This colour is then expressed in units of Red and Yellow.