Order Entry
Austria
ContactUsLinkComponent
Fume hood, moulded, large, SP Bel-Art
Fume hood, moulded, large, SP Bel-Art
Catalog # BELAH500000003
undefined
Fume hood, moulded, large, SP Bel-Art
Catalog # BELAH500000003

Specifications

  • W×D×H ext.:
    1070×510×760 mm
  • Description:
    Benchtop, front opening 880×360 mm
  • Pk:
    1

Specifications

About this item

PE hood, clear acrylic window.

  • Hood has an acrylic panel that slides up and down on the inner tracks
  • When the door is in closed position, a slight opening remains to facilitate airflow
  • Exhaust stack with outside diameter of 150 mm.

Shipping weight: 25,5 kg