Nitrile gloves, TouchNTuff® 93-250
: Ansell
These nitrile gloves resist a variety of industrial chemicals for longer periods than other nitrile disposable gloves. They also offer high tensile strength and 3 times the puncture resistance of latex or PVC gloves.
- Non sterile, powder-free, textured fingertips
- Ansell Grip Technology™ minimises the force required to grip dry, wet or oily objects, reducing hand and arm fatigue
- Proprietary nitrile formulation provides comfort for long wear periods
- Contains no natural rubber proteins, therefore protects against Type I skin allergies
- No transfer of silicone contaminants to contact surface
Ideal for use in food processing, laboratory applications, janitorial services, metal fabrication, and for automotive and transportation.
: EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS, Cat. III; AQL 1,5
Food contact approved
Food contact approved
- Catalog No:
- 112-4231
- 112-4776
- 112-4230
- 112-4232
- 112-4233
- Model:
- 93-250
- 93-250
- 93-250
- 93-250
- 93-250
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- Anti-static:
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
Frequently Bought Together