ContactUsLinkComponent
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Artikelnr. AXYGFDSS-1536-BK-R
Lieferant:  Corning
   Low Manufacturing Impact
undefined
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Artikelnr. AXYGFDSS-1536-BK-R
Lieferant:  Corning
   Low Manufacturing Impact
Spezifikationen
  • Packed:
    384 tips/rack, 10 racks/pack, 5 packs/case
  • Sterile:
    -
  • Volume:
    50 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Tip colour:
    Black
  • For use with:
    Head FDSS 7000
  • Filter:
    -
  • Pk:
    19.200
Spezifikationen
Über diesen Artikel
These black automation tips made of PP are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Conductive tips
  • Certified DNase-/RNase-free and non pyrogenic for complete protection against contamination
  • Optimised fine point for accurate dispensing