ContactUsLinkComponent
Pipette stand, rotary, SP Bel-Art
Pipette stand, rotary, SP Bel-Art
  BELAF189570000
undefined
Pipette stand, rotary, SP Bel-Art
  BELAF189570000
  • Ø×H:
    230×480 mm
  • For:
    94 pipettes
  • Colour:
    Blue/grey
  • Pk:
    1
Rotary pipette stand made of PP and PVC. Holds up to 94 pipettes and rotates on a central vertical axis for convenient selection.
  • Rotating disks are 18 cm (7⅛") in diameter
  • Lower disc consists of four concentric troughs with tapered sides and open bottoms which cradle the tips protectively
  • Upper rotating disc has a grid pattern concentric with the bottom disc keeping the pipettes vertical
  • Liquid draining off the pipette tip is retained on top of the stationary base by the outer rim
  • Low center of gravity stabilises the 48 cm high stand on the 23 cm diameter base