Express Eingabe
Austria
ContactUsLinkComponent
Transfer pipettes, Transpette™, SP Bel-Art
Transfer pipettes, Transpette™, SP Bel-Art
Catalog # 710-0083
undefined
Transfer pipettes, Transpette™, SP Bel-Art
Catalog # 710-0083
Specifications
  • Verpackt:
    Individually wrapped, 100/box
  • Steril:
    +
  • Kapazität:
    8×600 µl
  • Seq:
    02
  • VE:
    100
Specifications
About this item
Economical, single-use, 8-channel transfer pipette made of translucent plastic. Ideal for washing plates, feeding and supernatant aspiration from microwell plates.
  • No expensive pipettes and pipette tips needed
  • Should be disposed directly after 'hot' uses in order to avoid radiation contamination of costly pipettes
  • Available sterile or non sterile; can't be steam sterilised
  • Capacity: 600 µl per channel