ContactUsLinkComponent
Washable cleanroom floor mats, WorkZone Premier
Washable cleanroom floor mats, WorkZone Premier
Catalog # DYCECZ01/WZP/2GRAN
Supplier:  DYCEM
Washable cleanroom floor mats, WorkZone Premier
Catalog # DYCECZ01/WZP/2GRAN
Supplier:  DYCEM
Specifications
  • Colour:
    Grey
  • Seq:
    02
  • Pk:
    2 m2
Specifications
About this item
These mats are specially designed to remove contamination from areas subject to heavy wheeled traffic. Ideal for use at entrances and exits, corridors, packaging, warehouse and loading areas.
  • Attracts, collects and retains contamination from the wheels of forklifts, pallet trucks, large trolleys and carts
  • Prevents particles and microbes from getting into critical areas
Colour: Dark grey or grey