Order Entry
Austria
ContactUsLinkComponent
Stand alone Prep scale vacuum degassing system, Systec®
Stand alone Prep scale vacuum degassing system, Systec®
Catalog # HPLC0001-6484
Stand alone Prep scale vacuum degassing system, Systec®
Catalog # HPLC0001-6484

Specifications

  • Flow rate:
    40 ml/min
  • Max. pressure:
    4,8 bar
  • Description:
    Stand alone Prep scale vacuum degassing system
  • Int. volume:
    480 µl
  • Channels:
    2 × 13,8 ml
  • Pk:
    1

Specifications

About this item

The Systec® Stand-Alone Prep-Scale HPLC vacuum degassing systems is high-efficiency, in-line modules that remove dissolved gases from the mobile phase. Their unique design assures reliable continuous operation and the highest level of performance available without the need for helium sparging. Up to five solvent lines may be degassed simultaneously by one unit.