Orders LinkContactUsLinkComponent
Cold trap, type CP
Cold trap, type CP
Cold trap, CP S-29-O, borosilicate glass 3.3, with pressure guage GL 18 glass thread and screw-on cap, 1100 ml condensate capacity
Catalog # KGWA1707
Supplier:   KGW
Cold trap, type CP
Catalog # KGWA1707
Supplier:  KGW
Cold trap, type CP
Cold trap, type CP
Cold trap, CP S-29-O, borosilicate glass 3.3, with pressure guage GL 18 glass thread and screw-on cap, 1100 ml condensate capacity
Price
5 860,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents and these cold traps can be used for many different tasks. The most common application is gas cleaning, drying and condensation process. Model CP 1 has got an additional vacuum flange NW 16 with a vacuum gauge.
  • Compact design
  • Easy handling
  • Frame work on wheels
  • Suitable for aggressive mediums
  • Table board made of stainless steel
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
Specifications
  • Description:
    Cold trap, CP S-29-O, borosilicate glass 3.3, with pressure guage GL 18 glass thread and screw-on cap, 1100 ml condensate capacity