ContactUsLinkComponent
Beakers, tall form, super duty
Beakers, tall form, super duty
Catalog # 214-0404
Supplier:  DWK Life Sciences
Supplier #: 211182908
undefined
Beakers, tall form, super duty
Catalog # 214-0404
Supplier:  DWK Life Sciences
Supplier #: 211182908
Log in for availability
Select an Option
54 mm
Select an Option
150 ml
Select an Option
95 mm
Select an Option
1
About this item
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Higher mechanical stability compared with standard glass beakers due to reinforced sturdy rim
  • Even distribution of wall thickness
  • Very good chemical resistance, good thermal resistance
With easily legible graduation and labelling area. It is recommended to heat the Super Duty glass beakers slowly and evenly to prevent thermal stresses in the glass. For work at very high temperatures or with fast changes in temperature, the standard DURAN® beakers should be used, as these offer excellent resistance to temperature changes. However, mechanical stability is limited with these products in comparison with the Super Duty beakers.
With printed retrace code (batch identification).
Frequently Bought Together