Beaker tongs, stainless steel/rubber
Catalog # RSGA043.290
Supplier: RSG
Specifications
- Length:290 mm
- Pk:1
Specifications
About this item
Stainless steel 18/10. All-purpose beaker tongs with rubber protected tips. With second diameter for holding small flasks and beakers.