Storage cabinets, tall
Tall cabinets provide especially large storage space.
- Lockable doors
- Colour: Arctic white
Galvanised epoxy powder coated steel.
Frequently Bought Together
Tall cabinets provide especially large storage space.
Galvanised epoxy powder coated steel.