Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
GC ferrules, without holes
GC ferrules, without holes
GC ferrules, without holes
VARICR2121XX
:
VARIAN
GC ferrules, without holes
VARICR2121XX
:
VARIAN
Description:
Ferrules, vespel, blank, without holes, O.Ø
1
/
16
"
Pk:
10